missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD79b Monoclonal Antibody (C8), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551151
This item is not returnable.
View return policy
Description
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
Specifications
| CD79b | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P40259 | |
| Cd79b | |
| Recombinant Human CD79B protein, Asn37-Pro226 (Accession #P40259). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C8 | |
| Unconjugated | |
| Cd79b | |
| AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100511898, 974 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction