missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79B Antibody (HM79-11), Alexa Fluor™ 750, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NB100-65611AF750
This item is not returnable.
View return policy
Description
CD79B Monoclonal specifically detects CD79B in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD79B | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD79B | |
| CD79 alpha/CD79 beta heterodimer purified from WEH1231 B cells. | |
| Immunology | |
| 974 | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| HM79-11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
| Armenian Hamster | |
| 0.1 mL | |
| Primary | |
| NB100-65611 recognizes murine CD79 beta, which is expressed by B cells as part of the B cell receptor complex (immunoglobulin and the CD79 alpha/beta heterodimer). Murine CD79 beta is expressed at the cell surface of pro-B cells prior to surface immunoglobulin, and is expressed throughout B cell differentiation. CD79 beta is a B cell specific marker, valuable for the detection of B cells at all maturation stages. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction