missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79A Antibody (IGA/515), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47839MFV500
This item is not returnable.
View return policy
Description
CD79A Monoclonal antibody specifically detects CD79A in Human samples. It is validated for Flow Cytometry
Specifications
| CD79A | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| IGA/515 | |
| Flow Cytometry | |
| CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein | |
| Recombinant full-length human CD79A protein | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology, Phospho Specific | |
| 973 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction