missing translation for 'onlineSavingsMsg'
Learn More

CD77 Synthase Antibody (3E12), Novus Biologicals™

Product Code. 18322369 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322369 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322369 Supplier Novus Biologicals Supplier No. H00053947M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD77 Synthase Monoclonal antibody specifically detects CD77 Synthase in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD77 Synthase
Applications ELISA
Classification Monoclonal
Clone 3E12
Conjugate Unconjugated
Dilution ELISA
Formulation In 1x PBS, pH 7.4
Gene Alias A14GALTalpha 1,4-galactosyltransferase (globotriaosylceramide synthase, P blood group), A4GALT1, alpha 1,4-galactosyltransferase, Alpha-1,4-galactosyltransferase, Alpha-1,4-N-acetylglucosaminyltransferase, alpha4Gal-T1, CD77 synthase, EC 2.4.1.228, Gb3 synthase, Gb3S, Globotriaosylceramide synthase, lactosylceramide 4-alpha-galactosyltransferase, P blood group (P one antigen), P(k), P(k) antigen synthase, P1, P1/Pk synthase, PK, UDP-galactose:beta-D-galactosyl-beta1-R 4-alpha-D-galactosyltransferase
Host Species Mouse
Immunogen A4GALT (NP_059132, 254 a.a. ~ 353 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LTRVFKKWCSIRSLAESRACRGVTTLPPEAFYPIPWQDWKKYFEDINPEELPRLLSATYAVHVWNKKSQGTRFEATSRALLAQLHARYCPTTHEAMKMYL
Purification Method Ascites
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53947
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.