Learn More
Invitrogen™ CD75 Monoclonal Antibody (LN-1), eFluor™ 660, eBioscience™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 50-0759-42
Description
Description: This LN-1 monoclonal antibody reacts with human CD75, a cell surface carbohydrate determinant produced by alpha-2,6-sialytransferase. CD75 is expressed predominantly on mature B cells in peripheral blood and germinal centers, as well as several B cell lines (e.g., Raji, Ramos). CD75 is also expressed on non-immune tissues such as breast, prostate, colon, liver, and kidney. Reports demonstrate that CD75 is a ligand of CD22. Studies have shown that, depending on fixation and tissue processing, the LN-1 antibody also recognizes astrocytes and microglia in the human brain. Applications Reported: This LN-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This LN-1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein, which is normally found in the Golgi but which can be proteolytically processed to a soluble form, is involved in the generation of the cell-surface carbohydrate determinants and differentiation antigens HB-6, CDw75, and CD76. This protein is a member of glycosyltransferase family 29. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| CD75 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P15907 | |
| ST6GAL1 | |
| Affinity chromatography | |
| RUO | |
| 6480 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| LN-1 | |
| eFluor 660 | |
| ST6GAL1 | |
| Alpha 2,6-ST 1; AW742324; B-cell antigen CD75; beta galactoside alpha 2,6 sialyltransferase 1; beta-galactoside alpha-2,6-sialyltransferase 1; CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1; Sialyltransferase 1; sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase); Sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase); Sialyltransferase 1 (beta-galactoside alpha-26-sialytransferase); SIAT1; ST6 beta-galactosamide alpha-2,6-sialyltranferase 1; ST6 beta-galactoside alpha-2,6-sialyltransferase 1; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; St6gal; ST6Gal I; St6gal1; ST6GalI; St6Gal-I; ST6N | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.