missing translation for 'onlineSavingsMsg'
Learn More

CD75/CD75s Mouse anti-Human, Alexa Fluor 647, Clone: ZB55, BD

Product Code. 15884969
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15884969 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15884969 Supplier BD Biosciences Supplier No. 566350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The ZB55 monoclonal antibody specifically recognizes the carbohydrate CD75 (lactosamines) and CD75s (sialylated lactosamines) antigens which are expressed in association with various glycolipids and glycoproteins. CD75 is expressed on mature B cells and germinal center B lymphocytes whereas CD75s is expressed on mature B cells. CD75 expression is lost on B cells differentiating to plasma cells whereas CD75s expression is lost on both germinal center B cells and B cells differentiating into plasma cells. CD75 and CD75s may be expressed by some T cells and can be useful markers for distinguishing different types of malignant B cells.

Alexa Fluor™ 647 conjugates are highly photostable and remain fluorescent over a broad pH range. The excitation and emission maxima are nearly identical to those of APC. However, APC tends to be brighter while Alexa Fluor™ 647 is more optimal for intracellular applications. This fluorochrome exhibits uncommon photostability, making it an ideal choice for use in fluorescence microscopy. Due to nearly identical excitation and emission properties but different spillover characteristics, APC and Alexa Fluor™ 647 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD75/CD75s
Classification Monoclonal
Clone ZB55
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Gene Alias CD75/CD75s, CDw75
Host Species Mouse
Immunogen Human CD32 Peptides
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.