missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD74 Rabbit anti-Human, Mouse, Baboon, Clone: CLIP/3127R, Novus Biologicals™
Rabbit Monoclonal Antibody
£235.00
Specifications
| Antigen | CD74 |
|---|---|
| Clone | CLIP/3127R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
Description
CD74 Monoclonal specifically detects CD74 in Human, Mouse, Baboon samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
| CD74 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| 10 mM PBS with 0.05% BSA | |
| 972 | |
| IgG | |
| Protein A or G purified |
| CLIP/3127R | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Baboon | |
| CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
| SU-DHL-4 lymphoma cells | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title