missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD74 Mouse anti-Human, Clone: 2D8A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66390-1-IG-150UL
This item is not returnable.
View return policy
Description
Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.Specifications
| CD74 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04233 | |
| CD74 | |
| CD74 Fusion Protein Ag25503 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 2D8A1 | |
| Unconjugated | |
| CD74 | |
| CD74, DHLAG, HLADG, Ia GAMMA, Ii, p33 | |
| Mouse | |
| Protein A | |
| RUO | |
| 972 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction