missing translation for 'onlineSavingsMsg'
Learn More

CD73, APC, clone: AD2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0739-42

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13298219

  • £193.08 / Each


EU Stock 4
Add to basket

Description

Description

Description: This AD2 monoclonal antibody reacts with human CD73, a 5'-ectonucleotidase that converts 5'-adenosine monophosphate to adenosine. CD73 is expressed on the surface of endothelial cells, as well as B and T cells, including some CD4+Foxp3+ regulatory T cells. Adenosine production by these cells has been linked to the inhibition of CD4 T cell effector functions such as proliferation and cytokine secretion.Applications Reported: This AD2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This AD2 antibody has been pre-titrated and tested by flow cytometric analysis on normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD73
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P21589
CD73, E5NT, NT, NT5, NTE, eN, eNT, 5'-NT, 5'-nucleotidase, Purine 5-Prime-Nucleotidase, ecto-5'-nucleotidase
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
AD2
APC
NT5E
Conjugated
NT5E
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
4907
Videos

For Research Use Only.