missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD72 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13469-1-AP-150UL
This item is not returnable.
View return policy
Description
CD72 is a transmembrane glycoprotein expressed as a homodimer especially in B cells, but also in other antigen presenting cells such as dendritic cells and macrophages. Through one of its immunoreceptor tyrosine-based inhibitory motives (ITIMs), CD72 interacts with tyrosine phosphatase SHP-1, thereby suppressing B cell responsiveness. Binding of CD72 with its ligand CD100 (Sema4D) prevents BCR association and phosphorylation of CD72 and results in dissociation of SHP-1 from CD72, thus enables B cell activation.Specifications
| CD72 | |
| Polyclonal | |
| Unconjugated | |
| CD72 | |
| CD72, CD72 molecule, CD72b, Lyb 2, LYB2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12517, 971 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21854, P21855 | |
| Cd72 | |
| CD72 Fusion Protein Ag4262 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction