missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD72 Monoclonal Antibody (10.1.D2), PE

Description
CD72 Monoclonal Antibody for Flow
CD72 is a transmembrane glycoprotein expressed as a homodimer especially in B cells, but also in other antigen presenting cells such as dendritic cells and macrophages. Through one of its immunoreceptor tyrosine-based inhibitory motives (ITIMs), CD72 interacts with tyrosine phosphatase SHP-1, thereby suppressing B cell responsiveness. Binding of CD72 with its ligand CD100 (Sema4D) prevents BCR association and phosphorylation of CD72 and results in dissociation of SHP-1 from CD72, thus enables B cell activation.
Specifications
Specifications
| Antigen | CD72 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 10.1.D2 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with <0.1% sodium azide; pH 7.5 |
| Gene | CD72 |
| Gene Accession No. | P21855 |
| Gene Alias | B-cell differentiation antigen CD72; CD72; CD72 antigen; Cd72 molecule; CD72b; CD72c; Ly-19; Ly32; Ly-32; LYB2; Lyb-2; Ly-m19; lymphocyte antigen 32; RP11-331F9.9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?