missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD71 (Transferrin Receptor) Mouse, anti-Human, FITC, Clone: OKT9, STEMCELL Technologies
Mouse Monoclonal Antibody
£199.80 - £422.55
Specifications
| Antigen | CD71 (Transferrin Receptor) |
|---|---|
| Clone | OKT9 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17149051
|
Stemcell Technologies
60106FI.1 |
25 Tests |
£199.80
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17139051
|
Stemcell Technologies
60106FI |
100 Tests |
£422.55
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The OKT9 antibody reacts with CD71 (transferrin receptor), an ∼180 - 190 kDa disulfide-bonded homodimer and type 2 transmembrane glycoprotein expressed at high levels on a broad range of actively proliferating cells, as well as reticulocytes, monocytes, macrophages and marrow stromal cells
- Surface levels are low on resting leukocytes but are upregulated on lymphocytes, monocytes and macrophages following antigen or mitogen stimulation
- Expression is lost during differentiation of reticulocytes into mature erythrocytes
- CD71 mediates uptake of transferrin–iron complexes through receptor-mediated endocytosis and recycling of the apotransferrin–receptor complex to the cell surface
- It has also served as a target for drug delivery
- The OKT9 antibody binds an epitope that reportedly overlaps with that of antibody clone CY1G4 but is distinct from the ligand binding site for transferrin
Specifications
| CD71 (Transferrin Receptor) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| FITC | |
| Human | |
| Primary |
| OKT9 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title