missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCK Recombinant Rabbit Monoclonal Antibody (5S7P2)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558193

 View more versions of this product

Product Code. 30284187

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MATPPKRSCP SFSASSEGTR IKKISIEGNI AAGKSTFVNI LKQLCEDWEV VPEPVARWCN VQSTQDEFEE LTMSQKNGGN VLQMMYEKPE RWSFTFQTYA CLSRIRAQLA SLNGKLKDAE KPVLFFERSV YSDRYIFASN LYESECMNET EWTIYQDWHD WMNNQFGQSL ELDGIIYLQA TPETCLHRIY LRGRNEEQGI PLEYLEKLHY KHESWLLHRT LKTNFDYLQE VPILTLDVNE DFKDKYESLV EKVKEFLSTL.

Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DCK
Recombinant Monoclonal
1.34 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P27707, P48769
DCK
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DCK (NP_000779.1).
20 μL
Primary
Human, Rat
Antibody
IgG
ELISA, Western Blot
5S7P2
Unconjugated
DCK
DCK; Deoxycytidine kinase; deoxynucleoside kinase
Rabbit
Protein A
RUO
1633, 79127
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.