Learn More
CD71 Rat anti-Mouse, FITC, Clone: C2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553266
Additional Details : Weight : 0.01000kg
Description
The C2 monoclonal antibody specifically binds to CD71, the transferrin receptor. CD71 is a disulfide-linked homodimer of 95-kDa subunits. CD71 mediates one of the cellular mechanisms for iron uptake, and its expression is regulated according to the cell’s iron requirements. It is expressed at high levels on developing erythroid cells, and it is upregulated after mitogenic activation of B or T lymphocytes. The C2 monoclonal antibody selectivity inhibits some types of T- and B-cell activation by down-regulation of transferrin receptor expression, but it does not block binding of transferrin.
Specifications
CD71 | |
Monoclonal | |
0.5mg/mL | |
Transferrin Receptor; TR; TfR; TfR1; Tfrc; Trfr; Mtvr-1 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
C2 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse cell line | |
0.5 mg | |
Immunology | |
Mouse | |
IgG1 κ |
For Research Use Only.