Learn More
CD71 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30574-1-AP-150UL
 View more versions of this product
Product Code. 18084505
  • £349.00 / 150µL
Description
The CD71 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD71. This antibody recognizes human antigen. The CD71 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Specifications
CD71
260 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
CD71, p90, sTfR, T9, TFR, TFR1, TFRC, TR, Transferrin receptor protein 1, TRFR
150 μL
7037
Store at -20°C. Stable for one year after shipment.