Learn More
CD71 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30574-1-AP-20UL
This item is not returnable.
View return policy
Description
The CD71 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD71. This antibody recognizes human antigen. The CD71 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
| CD71 | |
| 260 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
| CD71, p90, sTfR, T9, TFR, TFR1, TFRC, TR, Transferrin receptor protein 1, TRFR | |
| 20 μL | |
| 7037 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |