Learn More
CD70 Monoclonal Antibody (FR70), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£97.50 - £156.00
Specifications
Antigen | CD70 |
---|---|
Clone | FR70 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15208589
|
Affymetrix eBioscience
12-0701-81 |
50 μg |
£97.50
50µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15218589
|
Affymetrix eBioscience
12-0701-82 |
100 μg |
£156.00
100µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The FR70 monoclonal antibody reacts with mouse CD70, a 30-33 kDa type II transmembrane glycoprotein. CD70 is a member of the TNF family and is expressed by activated mouse T and B lymphocytes. Interaction of CD70 with CD27, its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation. Applications Reported: This FR70 antibody has been reported for use in flow cytometric analysis. Applications Tested: This FR70 antibody has been tested by flow cytometric analysis of LPS-stimulated mouse splenocytes. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.Specifications
CD70 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; tumor necrosis factor ligand superfamily member 7 | |
Cd70 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cd70 |
FR70 | |
Flow Cytometry | |
PE | |
Rat | |
Mouse | |
O55237 | |
21948 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.