missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD70 Polyclonal Antibody

Product Code. 12560708
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12560708 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12560708 Supplier Invitrogen™ Supplier No. PA521943

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD70
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Cd70
Gene Accession No. P32970
Gene Alias CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; tumor necrosis factor ligand superfamily member 7
Gene Symbols Cd70
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of human CD70. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 970
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.