Learn More
CD7 Mouse anti-Human, PE, Clone: M-T701, BD
Mouse Monoclonal Antibody
£117.70 - £187.38
Specifications
| Antigen | CD7 |
|---|---|
| Clone | M-T701 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15891479
|
BD Biosciences
561934 |
25 Tests |
£122.00 £117.70 / Each Save £4.30 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15858098
|
BD Biosciences
555361 |
100 Tests |
£194.00 £187.38 / Each Save £6.62 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The M-T701 monoclonal antibody specifically binds to CD7. CD7 is a 40 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin superfamily. The CD7 antigen is also known as Leu-9, TP41, Tp40, GP40, and T-cell leukemia antigen. It is expressed on thymocytes, T cells, pre-B cells, and NK cells. CD7 is present in reduced density on monocytic cells and cell lines. Functional studies demonstrate that crosslinking of the CD7 can induce transmembrane calcium flux in T cells.
Specifications
| CD7 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| M-T701 | |
| Monoclonal | |
| Mouse | |
| Human | |
| P-CLL and Jurkat Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.