missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD7 Monoclonal Antibody (eBio124-1D1 (124-1D1)), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£154.00 - £340.00
Specifications
| Antigen | CD7 |
|---|---|
| Clone | eBio124-1D1 (124-1D1) |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055466
|
Invitrogen™
H018T03R03-A |
100 Tests |
£340.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055383
|
Invitrogen™
H018T02R03-A |
25 Tests |
£154.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow CytometrySpecifications
| CD7 | |
| 0.8 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P09564 | |
| 924 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBio124-1D1 (124-1D1) | |
| Flow Cytometry | |
| NovaFluor Red 700 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD7 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title