Learn More
CD7 Monoclonal Antibody (eBio124-1D1 (124-1D1)), eFluor™ 450, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£71.25 - £222.00
Specifications
| Antigen | CD7 |
|---|---|
| Clone | eBio124-1D1 (124-1D1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | eFluor 450 |
Description
CD7 (gp40, Leu9) is a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxy-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation.
CD7 (gp40, Leu9) is a 40 kDa member of the immunoglobulin gene superfamily. CD7 contains N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxy-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. Further, CD7 is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation. Expression of CD7 is an important marker used in leukemia diagnostics.Specifications
| CD7 | |
| Flow Cytometry | |
| eFluor 450 | |
| Mouse | |
| Human | |
| 924 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD7 |
| eBio124-1D1 (124-1D1) | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P09564 | |
| CD7 | |
| Primary | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.