missing translation for 'onlineSavingsMsg'
Learn More

CD7 Antibody [PE], Novus Biologicals™

Product Code. 30736932 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736932 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736932 Supplier Novus Biologicals Supplier No. AF7579P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CD7 Polyclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD7
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry, CyTOF
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
Host Species Sheep
Immunogen Kidney embryonic cell line HEK293-derived recombinant human CD7, Ala26-Pro180, Accession # P09564
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 924
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.