missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD7 Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human CD7.
Specifications
Specifications
| Assay Range | 15.625 pg/mL |
| Assay Sensitivity | 15.63-1000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes an antibody specific for Human CD7. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein |
| Gene ID (Entrez) | 924 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction