Learn More
Invitrogen™ CD69 Monoclonal Antibody (H1.2F3), PE-eFluor™ 610, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
£150.00 - £343.00
Specifications
| Antigen | CD69 |
|---|---|
| Clone | H1.2F3 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17124083
|
Invitrogen™
61-0691-80 |
25 μg |
£150.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17134083
![]() |
Invitrogen™
61-0691-82 |
100 μg |
£343.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The H1.2F3 MAb reacts with mouse CD69, also known as very early activation antigen (VEA). CD69 is approximately 35 kDa and is expressed on the surface as a disulfide-linked dimer. While a small subset of lymphocytes in the thymus, spleen and lymph nodes express this antigen, activation of both T and B cells rapidly upregulates the surface expression of CD69, suggesting a role for CD69 in lymphocyte development and activation.
CD69 (AIM, Active Inducer Molecule) is a gp28/34 disulfide bonded homodimer with a molecular weight of 60 kDa under non-reducing conditions. CD69 contains one or two N linked oligosacaride and the molecule is present on activated platelets. In normal peripheral blood a variable percentage of cells express the CD69 antigen, and it is involved in lymphocyte signal transduction. Expression CD69 is induced upon activation of T lymphocytes, and may play a role in proliferation. Furthermore, the protein may act to transmit signals in natural killer cells and platelets. Alternative splicing results in multiple transcript variants of CD69. Diseases associated with CD69 dysfunction include coccidiodomycosis and asthma.Specifications
| CD69 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 5830438K24Rik; Activation inducer molecule; activation inducer molecule (AIM/CD69); AI452015; AIM; BL-AC/P26; CD69; CD69 antigen; CD69 antigen (p60, early T-cell activation antigen); CD69 molecule; CLEC2C; C-type lectin domain family 2 member C; C-type lectin domain family 2, member C; EA1; early activation antigen CD69; early lymphocyte activation antigen; early T-cell activation antigen p60; GP32/28; leukocyte surface antigen Leu-23; MLR-3; VEA; Very Early Activation Antigen | |
| CD69 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD69 |
| H1.2F3 | |
| Flow Cytometry | |
| PE-eFluor 610 | |
| Armenian Hamster | |
| Mouse | |
| P37217 | |
| 12515 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.