missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD69 Armenian Hamster, PE-Cy7, Clone: H1.2F3, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand: EnquireBio QAB90-PE7-25ug
Additional Details : Weight : 0.01000kg
Specifications
CD69 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd69 | |
Protein A/G Purified | |
RUO | |
12515 | |
IgG |
Flow Cytometry | |
H1.2F3 | |
PE-Cyanine7 | |
NP_001028294 | |
Armenian Hamster | |
25 μg | |
Primary | |
Store at 2-4°C. |