missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD69 Antibody (Genefrontier patent anti-CD69) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28703-1mg
This item is not returnable.
View return policy
Description
CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD69 | |
| Monoclonal | |
| Unconjugated | |
| Q07108 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genefrontier patent anti-CD69 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 | |
| CD69 | |
| 1 mg | |
| Innate Immunity | |
| 969 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction