missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD69 Antibody (FN50), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43387PE
This item is not returnable.
View return policy
Description
CD69 Monoclonal specifically detects CD69 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD69 | |
| Monoclonal | |
| PE | |
| Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| FN50 | |
| Flow Cytometry | |
| CD69 | |
| The immunogen for this antibody was CD69. | |
| 0.1 mL | |
| Innate Immunity | |
| 969 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction