missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD69 Antibody (FN50), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
£433.00
Specifications
| Antigen | CD69 |
|---|---|
| Clone | FN50 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000 |
| Applications | Flow Cytometry |
Description
CD69 Monoclonal specifically detects CD69 in Human samples. It is validated for Flow Cytometry.Specifications
| CD69 | |
| 0.5 mg/mL | |
| Flow Cytometry | |
| Biotin | |
| RUO | |
| Human | |
| Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 | |
| CD69 | |
| IgG1 κ | |
| Affinity Purified |
| FN50 | |
| Flow Cytometry 1:10-1:1000 | |
| Monoclonal | |
| Mouse | |
| Innate Immunity | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
| 969 | |
| The immunogen for this antibody was CD69. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title