missing translation for 'onlineSavingsMsg'
Learn More

CD68/SR-D1 Mouse anti-Human, Mouse, Rat, Clone: KP1, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 49 publications

£238.00

Specifications

Antigen CD68/SR-D1
Clone KP1
Concentration 1 mg/mL
Dilution Western Blot 2 μg/mL, Flow Cytometry, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200, Electron Microscopy
Applications Western Blot
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18508481
View Documents
Novus Biologicals
NB100-683-0.025mg
0.025 mg
£238.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, Dual RNAscope ISH-IHC.
Specifications

Specifications

CD68/SR-D1
1 mg/mL
Western Blot
Unconjugated
Mouse
Cell Biology, Cytokine Research, Immunology
PBS
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Subcellular fraction of human alveolar macrophages.
Primary
Store at 4°C. Do not freeze.
KP1
Western Blot 2 μg/mL, Flow Cytometry, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200, Electron Microscopy
Monoclonal
Purified
RUO
Human
P34810
968
IgG1 κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.