missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68/SR-D1 Antibody (FA-11) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80656
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Flow (Intracellular), CyTOF-ready, Single Cell Western.
Specifications
| CD68/SR-D1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:100-1:2000, Flow Cytometry : 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence : 1:10-1:500, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin 2 ug/ml, Immunohistochemistry-Frozen : 1:10-1:500, Functional reported in scientific literature (PMID 11085350), Flow (Intracellular), CyTOF-ready, Single Cell Western | |
| CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, CyTOF | |
| FA-11 | |
| Unconjugated | |
| PBS | |
| CD68 | |
| This CD68/SR-D1 Antibody (FA-11) was developed against purified ConA acceptor glycoproteins from the P815 cell line. | |
| 0.1 mg | |
| Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
| 968 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction