missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68/SR-D1 Antibody (ED1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-985PE
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human, Mouse, Rat, Bovine, Feline, Primate, Monkey samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| CD68/SR-D1 | |
| Monoclonal | |
| PE | |
| CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This CD68/SR-D1 Antibody (ED1) recognizes a single chain glycoprotein of 110 kDa that is expressed predominantly on the lysosomal membrane of myeloid cells. Weak cell surface expression also occurs. The antigen is expressed by the majority of tissue macrophages and weakly by peripheral blood granulocytes. Studies have shown that the antigen recognized by ED1 is the rat homologue of human CD68. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| ED1 | |
| Flow Cytometry | |
| CD68 | |
| This CD68/SR-D1 Antibody (ED1) was developed against rat spleen cells | |
| 0.125 mL | |
| Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
| 968 | |
| Human, Mouse, Rat, Bovine, Feline, Primate, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction