missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68/SR-D1 Antibody (ED1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
£177.00 - £471.00
Specifications
| Antigen | CD68/SR-D1 |
|---|---|
| Clone | ED1 |
| Concentration | 1.0 mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18221334
|
Novus Biologicals
NB600-985-0.025MG |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18766853
|
Novus Biologicals
NB600-985 |
0.125 mg |
£471.00
125µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human, Mouse, Rat, Bovine, Feline, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay, Flow (Intracellular).Specifications
| CD68/SR-D1 | |
| 1.0 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q4FZY1 | |
| 968 | |
| IgG1 | |
| Protein A or G purified |
| ED1 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Primate, Feline, Bovine, Mouse, Human, Rat, Monkey | |
| CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| CD68 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title