missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68 Mouse, Clone: KP1 + C68/684, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 968-MSM3-P1
This item is not returnable.
View return policy
Specifications
| CD68 | |
| Cytoplasmic | |
| KP1 + C68/684 | |
| Unconjugated | |
| NP_001035148 | |
| Mouse | |
| 110kDa | |
| Protein A/G Purified | |
| RUO | |
| 968 | |
| IgG |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| CD68 | |
| Subcellular fraction of human alveolar macrophages (KP1); Recombinant human CD68 protein (C68/684) | |
| Tonsil, Lymph Node or Spleen | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction