Learn More
Invitrogen™ CD68 Monoclonal Antibody (815CU17), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 14-0687-37
Description
This 815CU17 MAb reacts with human CD68, a 110 kDa, transmembrane glycoprotein that belongs to the sialomucin family and is closely related to the lysosomal-associated membrane proteins (LAMP) and scavenger receptor. CD68 is predominantly an intracellular protein, found mainly in the late endosomal compartment but can be detected in smaller amounts on the surface of mainly myeloid-derived cells: monocytes, macrophages, dendritic cells (and in some langerhans cells), neutrophils, basophils, mast cells, myeloid progenitor cells, and a subset of CD34+ hematopoietic bone marrow progenitor cells. The function has not been fully elucidated but based on homology and structure, CD68 may play a role in antigen processing or presentation and protection of the lysosomal membrane from hydrolytic enzymes. Reports have also shown expression in activated T cells, and about 40% of peripheral blood B lymphocytes and 50% of all B-ALL. This 815CU17 antibody does not cross-react with mouse or rat CD68. For flow cytometric analysis of human CD68, we recommend clone eBioY1/82A.
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.
Specifications
| CD68 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P34810 | |
| Cd68 | |
| Affinity chromatography | |
| RUO | |
| 968 | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 815CU17 | |
| Unconjugated | |
| Cd68 | |
| CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen | |
| Mouse | |
| 2 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.