missing translation for 'onlineSavingsMsg'
Learn More

CD66c, PE, clone: KOR-SA3544, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0667-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15536816

  • £162.21 / 100 tests


EU Stock 3
Add to basket

Description

Description

Description: This KOR-SA3544 monoclonal antibody reacts with human CD66c (also known as CEACAM6), which is one of the splice variants of CD66. CD66c is expressed on endothelial and epithelial cells, as well as on neutrophils and monocytes. Myeloma and cancer cells, such as B-cell acute lymphoblastic lymphomas, also express CD66c. Moreover, expression of this receptor can be induced on T cells, B cells, and CD16-negative NK cells. Like the other CD66 splice variants, CD66c is involved in cell adhesion and growth, neutrophil activation, and signaling. Moreover, expression of CD66c in leukemic cells has been correlated with BCR/ABL1 rearrangement. Finally, studies have shown that the CD66 variants interact with each other homo- and heterotypically.Applications Reported: This KOR-SA3544 antibody has been reported for use in flow cytometric analysis.Applications Tested: This KOR-SA3544 antibody has been pre-titrated and tested by flow cytometry analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD66c
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P40199
CD66c, CEAL, NCA, carcinoembryonic antigen-related cell adhesion molecule 6, non-specific crossreacting antigen, normal cross-reacting antigen
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
KOR-SA3544
PE
CEACAM6
Conjugated
CEACAM6
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
4680
Videos

For Research Use Only.