missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD65 Monoclonal Antibody (VIM8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544035
This item is not returnable.
View return policy
Description
This antibody recognizes human CD65, an asialo-fucoganglioside expressed on the surface of peripheral blood granulocytes (highly) and monocytes (weakly).
This type 2 fucosylated carbohydrate antigen is expressed on the surface of all myeloid cells during development. Moreover, CD65 is expressed highly on granulocytes and weakly on monocytes in peripheral blood. CD65 has been reported to be a ligand for CD62E (E-Selectin). However, the sialylated form of CD65 (designated CD65s) can be used as a marker for acute myeloid leukemia (AML) as its expression increases as CD34 decreases. Finally, CD65 is important for extravascular infiltration of AML cells.
Specifications
| CD65 | |
| Monoclonal | |
| 1 mg/mL | |
| TBS with 15mM sodium azide; pH 8 | |
| Mouse | |
| Sequential chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgM |
| Flow Cytometry | |
| VIM8 | |
| Unconjugated | |
| Fucoganglioside Type 2; Fucoganglioside Type II | |
| THP-1 cell line. | |
| 100 μg | |
| Primary | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction