missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD64 Recombinant Rabbit Monoclonal Antibody (005), FITC

Rabbit Recombinant Monoclonal Antibody
£107.00 - £216.00
Specifications
| Antigen | CD64 |
|---|---|
| Clone | 005 |
| Concentration | 0.1 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30534289
![]() |
Invitrogen™
MA560568 |
100 Tests |
£216.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30535622
|
Invitrogen™
MA560569 |
25 Tests |
£107.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD64 Recombinant Monoclonal Antibody for Flow
CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.Specifications
| CD64 | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| FITC | |
| Rabbit | |
| RUO | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2 | |
| FCGR1A | |
| Primary | |
| Protein A |
| 005 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Rat | |
| FCGR1A | |
| 295279 | |
| Recombinant Rat CD64/FCGR1 protein (Met1-Pro285). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title