missing translation for 'onlineSavingsMsg'
Learn More

CD64 Mouse anti-Human, FITC, Clone: 10.1, BD

Mouse Monoclonal Antibody

£61.23 - £156.31

Specifications

Antigen CD64
Clone 10.1
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15830649
View Documents
BD Biosciences
560970
25 Tests
ONLINE EXCLUSIVE PRICE
£64.20
£61.23 / Each
Save £2.97
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15878238
View Documents
BD Biosciences
555527
100 Tests
ONLINE EXCLUSIVE PRICE
£163.00
£156.31 / Each
Save £6.69
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 10.1 monoclonal antibody specifically binds to CD64, a 72 kDa type I transmembrane glycoprotein that is a high affinity receptor for human IgG (FcγRI), especially the IgG1 and IgG3 subclasses. CD64 is expressed on monocytes, macrophages, dendritic cells, granulocytes activated with interferon-gamma and early myeloid lineage cells. CD64 associates with a signaling FcRγ homodimer to form the functional high affinity FcγRI complex. CD64 functions in both innate and adaptive immune responses and mediates endocytosis, phagocytosis, antigen presentation, antibody-dependent cellular toxicity, cytokine release and superoxide generation.

Specifications

Specifications

CD64
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
10.1
Monoclonal
Mouse
Human
Human Rheumatoid synovial fluid cells and fibronectin-purified monocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.