Learn More
CD63 Mouse anti-Human, PE, Clone: H5C6, BD
Mouse Monoclonal Antibody
£74.50 - £268.36
Specifications
| Antigen | CD63 |
|---|---|
| Clone | H5C6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15801479
|
BD Biosciences
561925 |
25 Tests |
£78.15 £74.50 / Each Save £3.65 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15888588
|
BD Biosciences
556020 |
100 Tests |
£280.00 £268.36 / Each Save £11.64 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The H5C6 monoclonal antibody reacts with CD63. CD63 is a 53 kDa, type III lysosomal glycoprotein, expressed on activated platelets, monocytes and macrophages. This molecule is also referred to in the literature as LIMP, gp55, melanoma-associated antigen ME491, Pltgp40, LAMP-3 and is a member of the tetraspan transmembrane 4 superfamily (TM4SF). It is widely expressed on surface and in the cytoplasm of various hematopoietic (monocytes, macrophages) and non-hematopoietic cells (endothelium, fibroblasts, osteoclasts, smooth muscle).
Specifications
| CD63 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| H5C6 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human Splenic Adherent Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.