Learn More
CD63 Mouse anti-Human, FITC, Clone: H5C6, BD
Mouse Monoclonal Antibody
£74.50 - £187.38
Specifications
| Antigen | CD63 |
|---|---|
| Clone | H5C6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15891469
|
BD Biosciences
561924 |
25 Tests |
£78.15 £74.50 / Each Save £3.65 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15848868
|
BD Biosciences
557288 |
100 Tests |
£195.00 £187.38 / Each Save £7.62 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The H5C6 monoclonal antibody reacts with CD63. CD63 is a 53 kDa, type III lysosomal glycoprotein, expressed on activated platelets, monocytes and macrophages. This molecule is also referred to in the literature as LIMP, gp55, melanoma-associated antigen ME491, Pltgp40, LAMP-3 and is a member of the tetraspan transmembrane 4 superfamily (TM4SF). It is widely expressed on surface and in the cytoplasm of various hematopoietic (monocytes, macrophages) and non-hematopoietic cells (endothelium, fibroblasts, osteoclasts, smooth muscle).
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD63 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| H5C6 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human Splenic Adherent Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.