missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD63 Monoclonal Antibody (H5C6), NovaFluor™ Red 700, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£160.00 - £340.00

Specifications

Antigen CD63
Clone H5C6
Concentration 0.8 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30056068
View Documents
Invitrogen™
H074T03R03-A
100 Tests
£340.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055183
View Documents
Invitrogen™
H074T02R03-A
25 Tests
£160.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD63
0.8 μg/Test
Monoclonal
Mouse
Human
P08962
967
Primary
4°C, store in dark, DO NOT FREEZE!
H5C6
Flow Cytometry
NovaFluor Red 700
RUO
PBS with 0.09% sodium azide; pH 7.2
C75951; CD 63; Cd63; CD63 antigen; CD63 antigen (melanoma 1 antigen); Cd63 molecule; CD63 protein; granulophysin; LAMP-3; Lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; mast cell antigen AD1; ME491; melanoma 1 antigen; melanoma-associated antigen ME491; melanoma-associated antigen MLA1; MLA1; Ocular melanoma-associated antigen; OMA81H; Tetraspanin-30; TSPAN30; tspan-30
IgG1 κ
Affinity chromatography
CD63
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.