Learn More
CD63 Monoclonal Antibody (H5C6), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 48-0639-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This H5C6 monoclonal antibody reacts with human CD63, a type III member of the tetraspanin family of transmembrane proteins. CD63 is expressed intracellularly on lysosomes, endosomes, and granules of resting platelets and basophils. However, cell surface expression of CD63 can be detected on activated basophils and platelets, monocytes, macrophages, and granulocytes. This receptor is also expressed on endothelial cells, fibroblasts, and smooth muscle cells. Studies have demonstrated that CD63 associates with integrins (VLA-3 and VLA-6) and TIMP-1 to mediate the allergic response. Applications Reported: This H5C6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This H5C6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (1 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD63 (LAMP-3, lysosome-associated membrane protein-3), a glycoprotein of tetraspanin family, is present in late endosomes, lysosomes and secretory vesicles of various cell types. CD63 is also present in the plasma membrane, usually following cell activation. Hence, CD63 has become a widely used basophil activation marker. In mast cells, however, CD63 exposition does not need their activation. CD63 interacts with integrins and affects phagocytosis and cell migration, it is also involved in H/K-ATPase trafficking regulation of ROMK1 channels. CD63 also serves as a T-cell costimulation molecule. Expression of CD63 can be used for predicting the prognosis in earlier stages of carcinomas. CD63 is expressed on activated platelets, and is a lysosomal membrane glycoprotein that is translocated to plasma membrane after platelet activation. CD63 is also present in monocytes and macrophages and is weakly expressed on granulocytes, B, and T cells. CD63 is identical to the melanoma-associated antigen which is ME491 and to the platelet antigen PTLGP40. Diseases associated with CD63 dysfunction include melanoma and Hermansky-Pudlak Syndrome.Specifications
CD63 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P08962 | |
CD63 | |
Affinity chromatography | |
RUO | |
967 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
H5C6 | |
eFluor 450 | |
CD63 | |
LAMP-3, ME491, MLA1, OMA81H, TSPAN30 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.