missing translation for 'onlineSavingsMsg'
Learn More

CD63 Antibody (SPM524), Allophycocyanin, Novus Biologicals™

Product Code. 18425042 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425042 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425042 Supplier Novus Biologicals Supplier No. NBP234779APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD63 Monoclonal antibody specifically detects CD63 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD63
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM524
Conjugate APC
Formulation PBS
Gene Alias CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30
Host Species Mouse
Immunogen Smooth plasma membrane fraction of MeWo cells
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis
Primary or Secondary Primary
Gene ID (Entrez) 967
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.