missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody (LAMP3/8705R), Novus Biologicals™
Rabbit Monoclonal Antibody
£224.00 - £403.00
Specifications
| Antigen | CD63 |
|---|---|
| Clone | LAMP3/8705R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650295
|
Novus Biologicals
NBP3-20500-20ug |
20 μg |
£224.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615134
|
Novus Biologicals
NBP3-20500-100ug |
100 μg |
£403.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD63 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
| 10mM PBS with 0.05% BSA | |
| 967 | |
| IgG κ | |
| Protein A or G purified |
| LAMP3/8705R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
| Recombinant full-length human CD63 protein (recognizes extracellular epitope) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title