Learn More
CD62P Rat anti-Mouse, FITC, Clone: RB40.34, BD
Rat Monoclonal Antibody
£126.17 - £442.55
Specifications
| Antigen | CD62P |
|---|---|
| Clone | RB40.34 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15881469
|
BD Biosciences
561923 |
0.1 mg |
£131.00 £126.17 / Each Save £4.83 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15847368
|
BD Biosciences
553744 |
0.5 mg |
£461.00 £442.55 / Each Save £18.45 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RB40.34 monoclonal antibody specifically binds to mouse P-selectin (CD62P), a 140 kDa protein which is expressed on activated platelets, activated endothelial cells, and megakaryocytes. P-selectin mediates the adhesion of neutrophils and monocytes to activated platelets and endothelial cells, mediates leukocyte rolling, and is involved in the migration of leukocytes into inflamed tissues. CD24 and CD162 (PSGL-1) are ligands of CD62P. mAb RB40.34 can block mouse P-selectin binding to its ligands in vitro and in vivo.
Specifications
| CD62P | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P-selectin-IgG1 Fusion | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| RB40.34 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 λ | |
| Affinity Purified | |
| P-selectin; Selp; LECAM3; LYAM3; PADGEM; GMP-140; Grmp |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.