Learn More
CD62L Mouse anti-Human, FITC, Clone: DREG56, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech FITC-65167-100TESTS
Description
The CD62L antibody from Proteintech is a mouse monoclonal antibody generated with pma-activated human peripheral blood leukocytes of human CD62L. This antibody recognizes human antigen. The CD62L antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
| CD62L | |
| Monoclonal | |
| 20 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| DREG56 | |
| FITC | |
| 100 Tests | |
| 6402 | |
| IgG1 |