Learn More
CD62L / L-Selectin Rat, PerCP-Cy5.5, Clone: MEL-14, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB49-PCP55-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15962118
  • £146.00 / 25µg
Estimated Shipment: 20-06-2024
to see stock.
Specifications
CD62L / L-Selectin
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2
Sell
Protein A/G Purified
RUO
20343
IgG2a κ
Flow Cytometry
MEL-14
PerCP-Cyanine5.5
NP_001157531
Rat
25 μg
Primary
Store at 2-4°C.