Learn More
Invitrogen™ CD62L (L-Selectin) Monoclonal Antibody (SK11), FITC, eBioscience™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 11-0625-42
Description
Description: This SK11 monoclonal antibody reacts with human and non-human primate CD62L (also known as L-selectin and LECAM). CD62L is expressed on neutrophils, monocytes, and subsets of T, B, and NK cells, as well as endothelial cells. This cell surface adhesion molecule binds glycolsylated, fucosylated and sulfated sialylated glycoproteins, including CD34, glycam-1, and MadCAM-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues. The SK11 antibody has been reported to recognize chimpanzee, rhesus, cynomolgus monkey, olive baboon, and pigtailed macaque as well as pig. Applications Reported: This SK11 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SK11 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.
Specifications
| CD62L (L-Selectin) | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P14151, Q95198, Q95237 | |
| SELL | |
| Affinity chromatography | |
| RUO | |
| 102142895, 450191, 6402, 701419 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| SK11 | |
| FITC | |
| SELL | |
| A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Baboon, Chimpanzee, Cynomolgus Monkey, Human, Monkey, Rhesus Macaque | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.