missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD62L (L-Selectin) Monoclonal Antibody (DREG-56 (DREG56)), NovaFluor™ Blue 610-30S, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£158.00 - £351.00

Specifications

Antigen CD62L (L-Selectin)
Clone DREG-56 (DREG56)
Concentration 0.2 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055108
View Documents
Invitrogen™
H009T03B05-A
100 Tests
£351.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055157
View Documents
Invitrogen™
H009T02B05-A
25 Tests
£158.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD62L (L-Selectin) Monoclonal antibody specifically detects CD62L (L-Selectin) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD62L (L-Selectin)
0.2 μg/Test
Monoclonal
Mouse
Human
P14151
6402
Primary
4°C, store in dark, DO NOT FREEZE!
DREG-56 (DREG56)
Flow Cytometry
NovaFluor Blue 610-30S
RUO
PBS with 0.09% sodium azide; pH 7.2
A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1
IgG1 κ
Affinity chromatography
SELL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.