missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD62L Fab Streptamer™ Isolation Kit MB

Product Code. 15312086 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15312086

Brand: IBA Lifesciences 68000204

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CD62L Fab Streptamer Isolation Kit MB is for reversible cell isolation delivering label-free target cells.

  • Strep-Tactin™ Magnetic Microbeads for Fab Streptamers™; 2 vials for 5 x 108 cells each
  • CD62L Fab-Strep, human for isolation of T-cells; 2 vials for 5 x 108 cells
  • Biotin stock solution for Streptamer™ technology; 2 x 1 mL
  • Buffer IS, Streptamer™ Washing Buffer, 10x concentrated; 50 mL
  • Microbeads are intended for use with a permanent magnet for manual cell isolation; we recommend our StrepMan magnet for 2 x 15 mL and 2 x 50 mL Falcon tubes.
  • Principle of Selection: Isolation of human CD62L+ T cells is performed by positive selection.
  • PBMCs are labeled with magnetic CD62L Fab Streptamers™.
  • Labeled cells are isolated in a strong magnet (StrepMan, horizontal position) where they migrate toward the tube wall on the side of the magnet.
  • This CD62L positive cell fraction is collected and cells are liberated from all labeling reagents by addition of biotin in a strong magnet (StrepMan, upright position).
  • The magnetic Streptamers™ migrate toward the tube wall and the label-free cells remain in the supernatant.
  • Biotin is removed by washing.
  • The resulting cell preparation is highly enriched with CD62L+ T cells with a purity of more than 90%.
  • No depletion steps and no columns are needed.

Specifications

Cell Type PBMCs
Material Magnetic Microspheres Coated with Strep-Tactin
No. of Cells 1 x 109
Quantity 1 Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.